Assay Details
| Gene Symbol: STAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268295.2 | XP_008266517.1 | 10 - 11 | 1217 | 67 | |
| XM_008268296.2 | XP_008266518.1 | 10 - 11 | 1091 | 67 | ||
More Information
Set Membership: |
|

STAM
Genomic Map