Assay Details
| Gene Symbol: POLI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713542.3 | XP_002713588.1 | 4 - 5 | 595 | 74 | |
| XM_017344182.1 | XP_017199671.1 | 4 - 5 | 537 | 74 | ||
More Information
Set Membership: |
|

POLI
Genomic Map