Assay Details
| Gene Symbol: LOC100343709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710450.3 | XP_002710496.1 | 14 - 15 | 1298 | 68 | |
| XM_008255595.2 | XP_008253817.1 | 15 - 16 | 1334 | 68 | ||
| XM_008255596.2 | XP_008253818.1 | 15 - 16 | 1288 | 68 | ||
| XM_008255598.2 | XP_008253820.1 | 15 - 16 | 1319 | 68 | ||
| XM_017341361.1 | XP_017196850.1 | 15 - 16 | 1226 | 68 | ||
| XM_017341362.1 | XP_017196851.1 | 15 - 16 | 1123 | 68 | ||
More Information
Set Membership: |
|

LOC100343709
Genomic Map