Assay Details
| Gene Symbol: CEP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711741.3 | XP_002711787.1 | 2 - 3 | 283 | 85 | |
| XM_008257792.2 | XP_008256014.1 | 2 - 3 | 283 | 85 | ||
| XM_017342493.1 | XP_017197982.1 | 2 - 3 | 283 | 85 | ||
More Information
Set Membership: |
|

CEP20
Genomic Map