Assay Details
| Gene Symbol: GATM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717894.3 | XP_002717940.3 | 6 - 7 | 1029 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GATM| Gene Symbol: GATM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717894.3 | XP_002717940.3 | 6 - 7 | 1029 | 80 | |
Set Membership: |
|