Assay Details
| Gene Symbol: ANKRD37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709330.3 | XP_002709376.1 | 3 - 4 | 354 | 70 | |
| XM_008250640.2 | XP_008248862.2 | 3 - 4 | 823 | 70 | ||
| XM_008250648.2 | XP_008248870.2 | 3 - 4 | 823 | 70 | ||
More Information
Set Membership: |
|

ANKRD37
Genomic Map