Assay Details
| Gene Symbol: C17H14orf93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718074.3 | XP_002718120.1 | 5 - 6 | 1603 | 73 | |
| XM_008269352.2 | XP_008267574.1 | 5 - 6 | 1612 | 73 | ||
More Information
Set Membership: |
|

C17H14orf93
Genomic Map