Assay Details
| Gene Symbol: PRORP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269496.2 | XP_008267718.1 | 2 - 3 | 265 | 91 | |
| XM_008269498.2 | XP_008267720.1 | 2 - 3 | 270 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008269496.2, XM_008269498.2 transcripts.
Set Membership: |
|

PRORP
Genomic Map