Assay Details
| Gene Symbol: STK35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256210.2 | XP_008254432.1 | 2 - 3 | 1752 | 83 | |
| XM_008256211.2 | XP_008254433.1 | 2 - 3 | 667 | 83 | ||
More Information
Set Membership: |
|

STK35
Genomic Map