Assay Details
| Gene Symbol: GPR68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719600.3 | XP_002719646.1 | 2 - 3 | 645 | 73 | |
| XM_008272121.2 | XP_008270343.2 | 2 - 3 | 512 | 73 | ||
| XM_017349476.1 | XP_017204965.1 | 2 - 3 | 695 | 73 | ||
| XM_017349477.1 | XP_017204966.1 | 2 - 3 | 516 | 73 | ||
| XM_017349478.1 | XP_017204967.1 | 2 - 3 | 386 | 73 | ||
| XM_017349479.1 | XP_017204968.1 | 2 - 3 | 671 | 73 | ||
| XM_017349480.1 | XP_017204969.1 | 2 - 3 | 621 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002719600.3, XM_008272121.2, XM_017349476.1, XM_017349477.1, XM_017349478.1, XM_017349479.1, XM_017349480.1 transcripts.
Set Membership: |
|

GPR68
Genomic Map