Assay Details
| Gene Symbol: TMPRSS11F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267796.2 | XP_008266018.1 | 10 - 11 | 1372 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS11F| Gene Symbol: TMPRSS11F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267796.2 | XP_008266018.1 | 10 - 11 | 1372 | 111 | |
Set Membership: |
|