Assay Details
| Gene Symbol: PABIR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720311.3 | XP_002720357.1 | 2 - 3 | 1119 | 99 | |
| XM_017349948.1 | XP_017205437.1 | 1119 | 99 | |||
More Information
Set Membership: |
|

PABIR2
Genomic Map