Assay Details
Gene Symbol: FAAP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711510.3 | XP_002711556.1 | 3 - 4 | 715 | 69 | |
XM_008257268.2 | XP_008255490.1 | 3 - 4 | 628 | 69 | ||
XM_017342213.1 | XP_017197702.1 | 2 - 3 | 240 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
