Assay Details
| Gene Symbol: LOC100357673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348776.1 | XP_017204265.1 | 21 - 22 | 2340 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100357673| Gene Symbol: LOC100357673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348776.1 | XP_017204265.1 | 21 - 22 | 2340 | 71 | |
Set Membership: |
|