Assay Details
| Gene Symbol: LOC100357673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348776.1 | XP_017204265.1 | 2 - 3 | 249 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017348776.1 transcripts.
Set Membership: |
|

LOC100357673
Genomic Map