Assay Details
| Gene Symbol: FER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713959.3 | XP_002714005.2 | 9 - 10 | 1326 | 77 | |
| XM_017344635.1 | XP_017200124.1 | 9 - 10 | 1328 | 77 | ||
| XM_017344636.1 | XP_017200125.1 | 9 - 10 | 1332 | 77 | ||
| XM_017344637.1 | XP_017200126.1 | 9 - 10 | 1331 | 77 | ||
More Information
Set Membership: |
|

FER
Genomic Map