Assay Details
| Gene Symbol: RBKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709946.3 | XP_002709992.1 | 5 - 6 | 568 | 61 | |
| XM_008254584.2 | XP_008252806.1 | 5 - 6 | 536 | 61 | ||
| XM_017340850.1 | XP_017196339.1 | 5 - 6 | 569 | 61 | ||
More Information
Set Membership: |
|

RBKS
Genomic Map