Assay Details
| Gene Symbol: MMRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267595.2 | XP_008265817.1 | 4 - 5 | 1138 | 61 | |
| XM_017347406.1 | XP_017202895.1 | 4 - 5 | 1138 | 61 | ||
More Information
Set Membership: |
|

MMRN1
Genomic Map