Assay Details
| Gene Symbol: DYNLRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257646.2 | XP_008255868.1 | 2 - 3 | 187 | 85 | |
| XM_008257647.2 | XP_008255869.1 | 2 - 3 | 597 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008257647.2 transcripts.
Set Membership: |
|

DYNLRB2
Genomic Map