Assay Details
| Gene Symbol: B3GALNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346886.1 | XP_017202375.1 | 3 - 4 | 282 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017346886.1 transcripts.
Set Membership: |
|

B3GALNT1
Genomic Map