Assay Details
Gene Symbol: LOC100354826 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017350475.1 | XP_017205964.1 | 399 | 133 | ||
XM_017350476.1 | XP_017205965.1 | 399 | 133 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
