Assay Details
| Gene Symbol: OAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349609.1 | XP_017205098.1 | 3 - 4 | 766 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OAS1| Gene Symbol: OAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349609.1 | XP_017205098.1 | 3 - 4 | 766 | 62 | |
Set Membership: |
|