Assay Details
| Gene Symbol: LOC103351522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271138.2 | XP_008269360.1 | 3 - 4 | 329 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103351522| Gene Symbol: LOC103351522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271138.2 | XP_008269360.1 | 3 - 4 | 329 | 106 | |
Set Membership: |
|