Assay Details
| Gene Symbol: LOC100354436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714390.3 | XP_002714436.1 | 2 - 3 | 878 | 104 | |
| XM_017345197.1 | XP_017200686.1 | 2 - 3 | 889 | 104 | ||
More Information
Set Membership: |
|

LOC100354436
Genomic Map