Assay Details
| Gene Symbol: DDX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719484.3 | XP_002719530.1 | 5 - 6 | 672 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDX5| Gene Symbol: DDX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719484.3 | XP_002719530.1 | 5 - 6 | 672 | 66 | |
Set Membership: |
|