Assay Details
| Gene Symbol: SEPTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252170.2 | XP_008250392.1 | 438 | 108 | ||
| XM_008252171.2 | XP_008250393.1 | 240 | 108 | |||
More Information
Set Membership: |
|

SEPTIN2
Genomic Map