Assay Details
Gene Symbol: ICAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002722843.3 | XP_002722889.1 | 1134 | 71 | ||
XM_008250740.2 | XP_008248962.1 | 1134 | 71 |

More Information
Set Membership: |
![]() ![]() |
