Assay Details
| Gene Symbol: GRO-B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717021.3 | XP_002717067.2 | 3 - 4 | 584 | 77 | |
| GenBank mRNA | L19157.1 | - | 2 - 3 | 209 | 77 | |
More Information
Set Membership: |
|

GRO-B
Genomic Map