Assay Details
| Gene Symbol: PTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711960.3 | XP_002712006.1 | 3 - 4 | 689 | 64 | |
| XM_008258043.2 | XP_008256265.1 | 3 - 4 | 695 | 64 | ||
| XM_017342645.1 | XP_017198134.1 | 3 - 4 | 306 | 64 | ||
More Information
Set Membership: |
|

PTN
Genomic Map