Assay Details
| Gene Symbol: TIRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259401.2 | XP_008257623.2 | 3 - 4 | 1387 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TIRAP| Gene Symbol: TIRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259401.2 | XP_008257623.2 | 3 - 4 | 1387 | 69 | |
Set Membership: |
|