Assay Details
| Gene Symbol: MMP28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719072.3 | XP_002719118.1 | 2 - 3 | 380 | 63 | |
| XM_017349069.1 | XP_017204558.1 | 2 - 3 | 382 | 63 | ||
| XM_017349070.1 | XP_017204559.1 | 2 - 3 | 380 | 63 | ||
More Information
Set Membership: |
|

MMP28
Genomic Map