Assay Details
| Gene Symbol: DISC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268257.2 | XP_008266479.1 | 12 - 13 | 2499 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DISC1| Gene Symbol: DISC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268257.2 | XP_008266479.1 | 12 - 13 | 2499 | 86 | |
Set Membership: |
|