Assay Details
| Gene Symbol: XRCC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713951.3 | XP_002713997.1 | 4 - 5 | 652 | 61 | |
| XM_008261904.2 | XP_008260126.1 | 4 - 5 | 575 | 61 | ||
| XM_008261905.2 | XP_008260127.1 | 5 - 6 | 938 | 61 | ||
| XM_017344563.1 | XP_017200052.1 | 4 - 5 | 509 | 61 | ||
| XM_017344564.1 | XP_017200053.1 | 4 - 5 | 576 | 61 | ||
More Information
Set Membership: |
|

XRCC4
Genomic Map