Assay Details
| Gene Symbol: LOC100349682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347094.1 | XP_017202583.1 | 1 - 2 | 297 | 95 | |
| XM_017347095.1 | XP_017202584.1 | 1 - 2 | 297 | 95 | ||
More Information
Set Membership: |
|

LOC100349682
Genomic Map