Assay Details
| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719485.3 | XP_002719531.1 | 8 - 9 | 1261 | 71 | |
| XM_008271725.2 | XP_008269947.1 | 8 - 9 | 1297 | 71 | ||
More Information
Set Membership: |
|

KPNA2
Genomic Map