Assay Details
| Gene Symbol: RAB37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723577.3 | XP_002723623.1 | 725 | 72 | ||
| XM_002723578.3 | XP_002723624.1 | 477 | 72 | |||
| XM_017339690.1 | XP_017195179.1 | 852 | 72 | |||
| XM_017339691.1 | XP_017195180.1 | 375 | 72 | |||
More Information
Set Membership: |
|

RAB37
Genomic Map