Assay Details
| Gene Symbol: MRPL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714984.3 | XP_002715030.1 | 2 - 3 | 993 | 92 | |
| XM_017345472.1 | XP_017200961.1 | 2 - 3 | 993 | 92 | ||
More Information
Set Membership: |
|

MRPL18
Genomic Map