Assay Details
| Gene Symbol: POGLUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713017.3 | XP_002713063.1 | 3 - 4 | 807 | 92 | |
| XM_017343585.1 | XP_017199074.1 | 3 - 4 | 599 | 92 | ||
| XM_017343586.1 | XP_017199075.1 | 2 - 3 | 251 | 92 | ||
More Information
Set Membership: |
|

POGLUT2
Genomic Map