Assay Details
| Gene Symbol: AGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268269.2 | XP_008266491.1 | 2 - 3 | 967 | 63 | |
| XM_008268270.2 | XP_008266492.1 | 2 - 3 | 947 | 63 | ||
More Information
Set Membership: |
|

AGT
Genomic Map