Assay Details
| Gene Symbol: SORD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269076.2 | XP_008267298.2 | 3 - 4 | 373 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SORD| Gene Symbol: SORD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269076.2 | XP_008267298.2 | 3 - 4 | 373 | 59 | |
Set Membership: |
|