Assay Details
| Gene Symbol: SNRPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261131.1 | XP_008259353.1 | 3 - 4 | 421 | 85 | |
| XM_008261132.2 | XP_008259354.1 | 3 - 4 | 359 | 85 | ||
More Information
Set Membership: |
|

SNRPD1
Genomic Map