Assay Details
| Gene Symbol: C6H16orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257698.2 | XP_008255920.1 | 1 - 2 | 431 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C6H16orf72| Gene Symbol: C6H16orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257698.2 | XP_008255920.1 | 1 - 2 | 431 | 81 | |
Set Membership: |
|