Assay Details
| Gene Symbol: IRAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259507.2 | XP_008257729.1 | 19 - 20 | 1684 | 79 | |
| XM_008259509.2 | XP_008257731.1 | 19 - 20 | 1681 | 79 | ||
| XM_008259510.2 | XP_008257732.1 | 18 - 19 | 1489 | 79 | ||
| XM_008259512.2 | XP_008257734.1 | 17 - 18 | 1630 | 79 | ||
| XM_017343328.1 | XP_017198817.1 | 18 - 19 | 1715 | 79 | ||
| XM_017343329.1 | XP_017198818.1 | 18 - 19 | 1549 | 79 | ||
| XM_017343330.1 | XP_017198819.1 | 14 - 15 | 1116 | 79 | ||
More Information
Set Membership: |
|

IRAG2
Genomic Map