Assay Details
| Gene Symbol: IRAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259507.2 | XP_008257729.1 | 8 - 9 | 780 | 77 | |
| XM_008259509.2 | XP_008257731.1 | 8 - 9 | 777 | 77 | ||
| XM_008259510.2 | XP_008257732.1 | 7 - 8 | 585 | 77 | ||
| XM_008259512.2 | XP_008257734.1 | 6 - 7 | 726 | 77 | ||
| XM_017343328.1 | XP_017198817.1 | 7 - 8 | 811 | 77 | ||
| XM_017343329.1 | XP_017198818.1 | 8 - 9 | 780 | 77 | ||
| XM_017343330.1 | XP_017198819.1 | 3 - 4 | 212 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008259512.2, XM_017343330.1 transcripts.
Set Membership: |
|

IRAG2
Genomic Map