Assay Details
| Gene Symbol: IRAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259507.2 | XP_008257729.1 | 14 - 15 | 1246 | 98 | |
| XM_008259509.2 | XP_008257731.1 | 14 - 15 | 1243 | 98 | ||
| XM_008259510.2 | XP_008257732.1 | 13 - 14 | 1051 | 98 | ||
| XM_008259512.2 | XP_008257734.1 | 12 - 13 | 1192 | 98 | ||
| XM_017343328.1 | XP_017198817.1 | 13 - 14 | 1277 | 98 | ||
| XM_017343330.1 | XP_017198819.1 | 9 - 10 | 678 | 98 | ||
More Information
Set Membership: |
|

IRAG2
Genomic Map