Assay Details
| Gene Symbol: SEPTIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337757.1 | XP_017193246.1 | 183 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017337757.1 transcripts.
Set Membership: |
|

SEPTIN1
Genomic Map