Assay Details
| Gene Symbol: HMCES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260330.1 | XP_008258552.1 | 5 - 6 | 746 | 60 | |
| XM_008260332.2 | XP_008258554.1 | 5 - 6 | 711 | 60 | ||
| XM_008260333.2 | XP_008258555.1 | 5 - 6 | 729 | 60 | ||
More Information
Set Membership: |
|

HMCES
Genomic Map