Assay Details
| Gene Symbol: ZSCAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249012.2 | XP_008247234.1 | 110 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008249012.2 transcripts.
Set Membership: |
|

ZSCAN18
Genomic Map