Assay Details
| Gene Symbol: LARP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722134.3 | XP_002722180.1 | 1 | 355 | 76 | |
| XM_008249369.2 | XP_008247591.1 | 3321 | 76 | |||
| XM_008249370.2 | XP_008247592.1 | 3321 | 76 | |||
| XM_008249371.2 | XP_008247593.1 | 1 | 355 | 76 | ||
| XM_017338074.1 | XP_017193563.1 | 3328 | 76 | |||
More Information
Set Membership: |
|

LARP4B
Genomic Map