Assay Details
| Gene Symbol: CLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715424.3 | XP_002715470.1 | 11 - 12 | 1499 | 63 | |
| XM_008264345.2 | XP_008262567.1 | 11 - 12 | 1502 | 63 | ||
| XM_008264346.2 | XP_008262568.1 | 11 - 12 | 1499 | 63 | ||
| XM_008264347.2 | XP_008262569.1 | 11 - 12 | 1497 | 63 | ||
| XM_017345826.1 | XP_017201315.1 | 11 - 12 | 1524 | 63 | ||
| XM_017345827.1 | XP_017201316.1 | 11 - 12 | 1522 | 63 | ||
More Information
Set Membership: |
|

CLK2
Genomic Map